Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A1
Entrez Gene ID
1291
Gene Name
collagen type VI alpha 1
Gene Aliases
BTHLM1, OPLL, UCHMD1
Chromosome Location
Chr.21: 45981749 - 46005049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001848.2 NP_001839.2 27 - 28 1875 70
GenBank mRNA AK307989.1 - 27 - 28 1853 70
BC052575.1 - 27 - 28 1855 70

View Full Product Details