Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MARK2P9
Entrez Gene ID
100507674
Gene Name
microtubule affinity regulating kinase 2 pseudogene 9
Gene Aliases
-
Chromosome Location
Chr.10: 92418661 - 92419606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_038243.2 1 - 1 710 110


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream membrane associated ring-CH-type finger 5 MARCH5 MARCH-V, MITOL, RNF153 -64.7kb Sense Chr. - 92291163 - 92353964 on Build GRCh38
Downstream insulin degrading enzyme IDE INSULYSIN 32.1kb Anti-sense Chr. - 92451684 - 92574484 on Build GRCh38

View Full Product Details