Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DENND4C
- Entrez Gene ID
-
55667
- Gene Name
- DENN domain containing 4C
- Gene Aliases
-
C9orf55, C9orf55B, RAB10GEF, bA513M16.3
- Chromosome Location
-
Chr.9: 19230765 - 19374268 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073201.3 |
|
19 - 20 |
2687 |
76 |
GenBank mRNA |
AK299842.1 |
- |
6 - 7 |
742 |
76 |
BC144484.1 |
- |
16 - 17 |
2543 |
76 |
BC144485.1 |
- |
15 - 16 |
1919 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DC325436.1 |
- |
4 - 5 |
387 |
|
Chr. 9 - 19335084 - 19342706 [+] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
perilipin 2 |
PLIN2 |
ADFP, ADRP |
-210.0kb
|
Anti-sense |
Chr. - 19108391 - 19127606 on Build GRCh38 |
Downstream |
ribosomal protein S6 |
RPS6 |
S6 |
35.1kb
|
Anti-sense |
Chr. - 19376256 - 19380237 on Build GRCh38 |
View Full Product Details