The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003012.1 | 1 - 1 | 76 | 96 | |
GenBank mRNA | AJ609487.1 | - | 1 - 1 | 76 | 96 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | NOP2 nucleolar protein | NOP2 | NOL1, NOP120, NSUN1, p120 | -13.1kb | Sense | Chr. - 6556870 - 6568332 on Build GRCh38 |
Downstream | lysophosphatidic acid receptor 5 | LPAR5 | GPR92, GPR93, KPG_010, LPA5 | 37.2kb | Sense | Chr. - 6618835 - 6636131 on Build GRCh38 |