Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
Chromosome Location
Chr.11: 128685263 - 128813267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167681.2 NP_001161153.1 5 - 6 987 69
NM_001271010.1 NP_001257939.1 5 - 6 1104 69
NM_002017.4 NP_002008.2 4 - 5 933 69
XM_011542701.2 XP_011541003.1 4 - 5 748 69
XM_011542702.1 XP_011541004.1 4 - 5 1079 69
XM_017017405.1 XP_016872894.1 4 - 5 682 69
XM_017017406.1 XP_016872895.1 4 - 5 871 69
GenBank mRNA AK300153.1 - 5 - 6 811 69
AK313370.1 - 4 - 5 787 69
AY029368.1 - 4 - 5 590 69
BC001670.1 - 4 - 5 713 69
BC010115.1 - 4 - 5 715 69
M93255.1 - 5 - 6 934 69
M98833.3 - 4 - 5 762 69
S45205.1 - 4 - 5 723 69
X67001.1 - 4 - 5 732 69

View Full Product Details