Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL6
Entrez Gene ID
10402
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 6
Gene Aliases
SIAT10, ST3GALVI
Chromosome Location
Chr.3: 98732236 - 98795845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271142.1 NP_001258071.1 10 - 11 1290 174
NM_001271145.1 NP_001258074.1 9 - 10 1231 174
NM_001271146.1 NP_001258075.1 9 - 10 1220 174
NM_001271147.1 NP_001258076.1 9 - 10 1194 174
NM_001271148.1 NP_001258077.1 6 - 7 978 174
NM_001323352.1 NP_001310281.1 10 - 11 1392 174
NM_001323353.1 NP_001310282.1 8 - 9 1156 174
NM_001323358.1 NP_001310287.1 11 - 12 1401 174
NM_001323359.1 NP_001310288.1 10 - 11 1301 174
NM_001323360.1 NP_001310289.1 11 - 12 1462 174
NM_001323362.1 NP_001310291.1 9 - 10 1328 174
NM_001323363.1 NP_001310292.1 8 - 9 1167 174
NM_001323364.1 NP_001310293.1 9 - 10 1339 174
NM_001323365.1 NP_001310294.1 10 - 11 1231 174
NM_001323366.1 NP_001310295.1 11 - 12 1301 174
NM_001323367.1 NP_001310296.1 9 - 10 1048 174
NM_001323368.1 NP_001310297.1 9 - 10 1056 174
NM_006100.3 NP_006091.1 10 - 11 1403 174
GenBank mRNA AB022918.1 - 9 - 10 1170 174
AF119391.1 - 9 - 10 1214 174
AK001922.1 - 9 - 10 981 174
BC001266.2 - 10 - 11 1218 174
BC023312.1 - 10 - 11 1379 174

View Full Product Details