Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
Chromosome Location
Chr.11: 128685263 - 128813267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167681.2 NP_001161153.1 3 - 4 629 99
NM_002017.4 NP_002008.2 2 - 3 575 99
XM_011542701.2 XP_011541003.1 2 - 3 390 99
XM_011542702.1 XP_011541004.1 2 - 3 721 99
XM_017017405.1 XP_016872894.1 2 - 3 324 99
XM_017017406.1 XP_016872895.1 2 - 3 513 99
GenBank mRNA AF147318.1 - 1 - 2 93 99
AK300153.1 - 3 - 4 453 99
AK313370.1 - 2 - 3 429 99
AY029368.1 - 2 - 3 232 99
BC001670.1 - 2 - 3 355 99
BC010115.1 - 2 - 3 357 99
M98833.3 - 2 - 3 404 99
S45205.1 - 2 - 3 365 99
X67001.1 - 2 - 3 374 99

View Full Product Details