The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024766.4 | NP_079042.1 | 3 - 4 | 478 | 80 |
XM_011533111.2 | XP_011531413.1 | 3 - 4 | 444 | 80 | |
GenBank mRNA | AK027104.1 | - | 3 - 4 | 418 | 80 |
BC053733.1 | - | 3 - 4 | 405 | 80 |