The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205273.2 | NP_001192202.1 | 1 - 2 | 66 | 124 |
GenBank mRNA | FJ211597.1 | - | 1 - 2 | 66 | 124 |