The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258024.1 | NP_001244953.1 | 3 - 4 | 358 | 68 |
GenBank mRNA | AY669507.1 | - | 3 - 4 | 358 | 68 |