The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004342.3 | NP_001004342.3 | 6 - 7 | 1721 | 81 |
NM_001300889.1 | NP_001287818.1 | 8 - 9 | 1535 | 81 | |
XM_011544192.2 | XP_011542494.1 | 6 - 7 | 1715 | 81 | |
XM_017001323.1 | XP_016856812.1 | 6 - 7 | 1721 | 81 | |
GenBank mRNA | AY253917.1 | - | 8 - 9 | 1493 | 81 |