Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN1
Entrez Gene ID
79906
Gene Name
MORN repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 2321253 - 2391751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301060.1 NP_001287989.1 1 - 2 294 95
NM_024848.2 NP_079124.1 1 - 2 294 95
XM_011542169.1 XP_011540471.1 1 - 2 294 95
XM_011542170.2 XP_011540472.1 1 - 2 294 95
XM_011542173.2 XP_011540475.1 1 - 2 294 95
XM_011542174.2 XP_011540476.1 1 - 2 294 95
XM_011542176.1 XP_011540478.1 1 - 2 294 95
XM_011542177.1 XP_011540479.1 1 - 2 294 95
XM_017002365.1 XP_016857854.1 1 - 2 294 95
GenBank mRNA AK024003.1 - 1 - 2 97 95
BC021704.2 - 1 - 2 250 95
DB041436.1 - 1 - 2 294 95

View Full Product Details