Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRN3
Entrez Gene ID
54700
Gene Name
RRN3 homolog, RNA polymerase I transcription factor
Gene Aliases
A-270G1.2, TIFIA
Chromosome Location
Chr.16: 15060022 - 15094335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301064.1 NP_001287993.1 11 - 12 1144 111
NM_018427.4 NP_060897.3 12 - 13 1234 111
XM_005255376.1 XP_005255433.1 10 - 11 1009 111
XM_005255377.4 XP_005255434.1 9 - 10 958 111
XM_011522540.2 XP_011520842.1 7 - 8 646 111
XM_011522541.1 XP_011520843.1 11 - 12 1073 111
GenBank mRNA AF227156.1 - 12 - 13 1200 111
AJ272050.1 - 12 - 13 1154 111
AK225662.1 - 12 - 13 1210 111
AK295486.1 - 9 - 10 827 111
AK300253.1 - 11 - 12 1053 111
AK302994.1 - 9 - 10 881 111
AK303405.1 - 8 - 9 727 111
AK304856.1 - 12 - 13 1100 111
AK314769.1 - 12 - 13 1148 111
AK316520.1 - 11 - 12 1058 111
BC036182.1 - 12 - 13 1149 111
BC071868.1 - 4 - 5 469 111
BC132688.1 - 12 - 13 1151 111
BC132690.1 - 12 - 13 1151 111
DA764092.1 - 4 - 5 437 111

View Full Product Details