The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301716.1 | NP_001288645.1 | 2 - 3 | 382 | 84 |
NM_001301717.1 | NP_001288646.1 | 2 - 3 | 138 | 84 | |
NM_001301718.1 | NP_001288647.1 | 2 - 3 | 228 | 84 | |
NM_001838.3 | NP_001829.1 | 2 - 3 | 132 | 84 | |
GenBank mRNA | AK127810.1 | - | 2 - 3 | 120 | 84 |
AK310277.1 | - | 2 - 3 | 382 | 84 | |
AK312732.1 | - | 2 - 3 | 116 | 84 | |
AY587876.1 | - | 1 - 2 | 57 | 84 | |
BC035343.1 | - | 2 - 3 | 98 | 84 | |
DA814913.1 | - | 2 - 3 | 132 | 84 | |
HY017993.1 | - | 2 - 3 | 99 | 84 | |
HY021417.1 | - | 2 - 3 | 229 | 84 | |
L08176.1 | - | 2 - 3 | 120 | 84 | |
L31581.1 | - | 2 - 3 | 123 | 84 |