Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
Chromosome Location
Chr.12: 49622717 - 49644669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031698.2 NP_001026868.2 13 - 14 1347 78
NM_012272.2 NP_036404.1 12 - 13 1637 78
XM_006719324.3 XP_006719387.1 12 - 13 2401 78
XM_006719325.3 XP_006719388.1 12 - 13 2401 78
XM_011538135.2 XP_011536437.1 12 - 13 2401 78
XM_011538136.2 XP_011536438.1 12 - 13 2401 78
XM_011538137.2 XP_011536439.1 11 - 12 2320 78
XM_011538138.2 XP_011536440.1 11 - 12 2290 78
XM_011538139.2 XP_011536441.1 13 - 14 1341 78
XM_011538140.2 XP_011536442.1 13 - 14 1341 78
XM_011538141.1 XP_011536443.1 12 - 13 1167 78
XM_011538143.2 XP_011536445.1 12 - 13 1183 78
XM_011538144.2 XP_011536446.1 11 - 12 1056 78
XM_017019135.1 XP_016874624.1 12 - 13 1104 78
XM_017019136.1 XP_016874625.1 11 - 12 1056 78
XM_017019137.1 XP_016874626.1 11 - 12 1056 78
XM_017019138.1 XP_016874627.1 12 - 13 2401 78
XM_017019139.1 XP_016874628.1 8 - 9 1067 78
GenBank mRNA AL834216.1 - 12 - 13 1637 78
BC031917.1 - 10 - 11 943 78
BC050398.2 - 13 - 14 1183 78
BC067364.1 - 12 - 13 1168 78

View Full Product Details