The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127394.3 | NP_001120866.1 | 1 - 2 | 242 | 99 |
NM_001300764.1 | NP_001287693.1 | 1 - 2 | 242 | 99 | |
NM_001300766.1 | NP_001287695.1 | 1 - 2 | 242 | 99 | |
NM_052965.3 | NP_443197.1 | 1 - 2 | 242 | 99 | |
NR_023349.2 | 1 - 2 | 242 | 99 | ||
NR_125335.1 | 1 - 2 | 242 | 99 | ||
XM_006711148.1 | XP_006711211.1 | 1 - 2 | 191 | 99 | |
XM_011509139.2 | XP_011507441.1 | 1 - 2 | 1686 | 99 | |
XM_017000228.1 | XP_016855717.1 | 1 - 2 | 1686 | 99 | |
XM_017000229.1 | XP_016855718.1 | 1 - 2 | 1686 | 99 | |
XM_017000230.1 | XP_016855719.1 | 1 - 2 | 1686 | 99 | |
GenBank mRNA | AF288394.1 | - | 1 - 2 | 217 | 99 |
AK098196.1 | - | 1 - 2 | 140 | 99 | |
AK296655.1 | - | 1 - 2 | 137 | 99 | |
BC022030.1 | - | 1 - 2 | 187 | 99 | |
BI821111.1 | - | 1 - 2 | 167 | 99 | |
BM146150.1 | - | 1 - 2 | 244 | 99 | |
BM887891.1 | - | 1 - 2 | 74 | 99 | |
BU195337.1 | - | 1 - 2 | 214 | 99 |