Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSEN15
Entrez Gene ID
116461
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
C1orf19, PCH2F, sen15
Chromosome Location
Chr.1: 184050207 - 184095846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300764.1 NP_001287693.1 3 - 4 450 69
NM_001300766.1 NP_001287695.1 3 - 4 450 69
NM_052965.3 NP_443197.1 3 - 4 450 69
XM_006711148.1 XP_006711211.1 3 - 4 399 69
GenBank mRNA AF288394.1 - 3 - 4 425 69
AK098196.1 - 3 - 4 348 69
BC022030.1 - 3 - 4 395 69
BI821111.1 - 3 - 4 375 69
BP326115.1 - 3 - 4 375 69

View Full Product Details