The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085423.1 | NP_001078892.1 | 6 - 7 | 901 | 93 |
NM_001291316.1 | NP_001278245.1 | 5 - 6 | 616 | 93 | |
NM_001291317.1 | NP_001278246.1 | 5 - 6 | 631 | 93 | |
XM_017024486.1 | XP_016879975.1 | 6 - 7 | 994 | 93 | |
XM_017024487.1 | XP_016879976.1 | 6 - 7 | 914 | 93 | |
GenBank mRNA | AB542950.1 | - | 6 - 7 | 853 | 93 |
AB542951.1 | - | 5 - 6 | 562 | 93 | |
AB542952.1 | - | 5 - 6 | 577 | 93 | |
BC053534.1 | - | 5 - 6 | 795 | 93 | |
BC130406.1 | - | 6 - 7 | 876 | 93 | |
BC136594.1 | - | 6 - 7 | 876 | 93 | |
BC144272.1 | - | 5 - 6 | 591 | 93 | |
BE296352.1 | - | 3 - 4 | 402 | 93 |