The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104387.1 | 38 - 39 | 5641 | 83 | |
NR_104388.1 | 38 - 39 | 5632 | 83 | ||
NR_104389.1 | 37 - 38 | 5539 | 83 | ||
NR_104390.1 | 38 - 39 | 5545 | 83 | ||
NR_104391.1 | 39 - 40 | 5567 | 83 | ||
NR_104392.1 | 38 - 39 | 5561 | 83 | ||
NR_104393.1 | 39 - 40 | 5647 | 83 | ||
NR_104394.1 | 38 - 39 | 5518 | 83 | ||
NR_104395.1 | 38 - 39 | 5570 | 83 | ||
GenBank mRNA | AJ549248.3 | - | 13 - 14 | 1466 | 83 |
AJ549249.2 | - | 9 - 10 | 1003 | 83 | |
AJ549250.3 | - | 12 - 13 | 1364 | 83 | |
AJ555148.2 | - | 13 - 14 | 1370 | 83 | |
AJ555149.2 | - | 14 - 15 | 1392 | 83 | |
AM711892.1 | - | 13 - 14 | 1386 | 83 | |
AM711893.1 | - | 14 - 15 | 1472 | 83 | |
AM711894.1 | - | 13 - 14 | 1343 | 83 | |
AM711895.1 | - | 13 - 14 | 1395 | 83 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | inhibitor of growth family, X-linked (pseudogene) | INGX | ING1-like, ING2 | Anti-sense | Chr. - 71491681 - 71492449 on Build GRCh38 | |
Downstream | O-linked N-acetylglucosamine (GlcNAc) transferase | OGT | HINCUT-1, HRNT1, O-GLCNAC | 4.3kb | Sense | Chr. - 71533062 - 71575897 on Build GRCh38 |