Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR6776
- Entrez Gene ID
-
102465465
- Gene Name
- microRNA 6776
- Gene Aliases
-
hsa-mir-6776
- Chromosome Location
-
Chr.17: 2692861 - 2692919 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_106834.1 |
|
1 - 1 |
28 |
54 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
clustered mitochondria homolog |
CLUH |
CLU1, KIAA0664 |
|
Sense |
Chr. - 2689386 - 2711633 on Build GRCh38 |
Downstream |
uncharacterized LOC105371591 |
LOC105371591 |
|
13.6kb
|
Sense |
Chr. - 2706477 - 2708492 on Build GRCh38 |
View Full Product Details