Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERI3-IT1
Entrez Gene ID
100874278
Gene Name
ERI3 intronic transcript 1
Gene Aliases
-
Chromosome Location
Chr.1: 44222595 - 44244283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110056.1 2 - 3 204 104
GenBank mRNA DB453456.1 - 2 - 3 204 104


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ERI1 exoribonuclease family member 3 ERI3 PINT1, PRNPIP Sense Chr. - 44221070 - 44355298 on Build GRCh38
Downstream ring finger protein 220 RNF220 C1orf164 162.7kb Anti-sense Chr. - 44404846 - 44651724 on Build GRCh38

View Full Product Details