Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C17orf80
- Entrez Gene ID
-
55028
- Gene Name
- chromosome 17 open reading frame 80
- Gene Aliases
-
HLC-8, MIG3, SPEP1
- Chromosome Location
-
Chr.17: 73232233 - 73248959 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110105.1 |
|
2 - 3 |
197 |
72 |
GenBank mRNA |
BG754800.1 |
- |
1 - 2 |
114 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE883983.1 |
- |
1 - 2 |
96 |
|
Chr. 17 - 73233226 - 73247455 [+] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cleavage and polyadenylation specific factor 4 like |
CPSF4L |
|
|
Anti-sense |
Chr. - 73242343 - 73263778 on Build GRCh38 |
Downstream |
CDC42 effector protein 4 |
CDC42EP4 |
BORG4, CEP4, KAIA1777 |
41.3kb
|
Anti-sense |
Chr. - 73283624 - 73312004 on Build GRCh38 |
View Full Product Details