Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LIN54
- Entrez Gene ID
-
132660
- Gene Name
- lin-54 DREAM MuvB core complex component
- Gene Aliases
-
CXCDC1, JC8.6, MIP120, TCX1
- Chromosome Location
-
Chr.4: 82924603 - 83012951 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110253.1 |
|
2 - 3 |
1065 |
85 |
GenBank mRNA |
BC109277.2 |
- |
2 - 3 |
941 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
THAP domain containing 9 |
THAP9 |
hTh9 |
-50.0kb
|
Anti-sense |
Chr. - 82900650 - 82920283 on Build GRCh38 |
Downstream |
COP9 signalosome subunit 4 |
COPS4 |
CSN4 |
50.2kb
|
Anti-sense |
Chr. - 83035086 - 83075818 on Build GRCh38 |
View Full Product Details