Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ELFN2
- Entrez Gene ID
-
114794
- Gene Name
- extracellular leucine rich repeat and fibronectin type III domain containing 2
- Gene Aliases
-
LRRC62, PPP1R29
- Chromosome Location
-
Chr.22: 37340644 - 37427470 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110512.1 |
|
1 - 2 |
175 |
85 |
GenBank mRNA |
DA365834.1 |
- |
1 - 2 |
150 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100506271 |
LOC100506271 |
|
|
Sense |
Chr. - 37352190 - 37354839 on Build GRCh38 |
Downstream |
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase |
MFNG |
|
41.6kb
|
Sense |
Chr. - 37469063 - 37486440 on Build GRCh38 |
View Full Product Details