Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C22orf34
- Entrez Gene ID
-
348645
- Gene Name
- chromosome 22 open reading frame 34
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 49414524 - 49657542 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110523.1 |
|
2 - 3 |
479 |
123 |
GenBank mRNA |
BC033837.1 |
- |
2 - 3 |
452 |
123 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF797797.1 |
- |
2 - 3 |
438 |
|
Chr. 22 - 49415917 - 49657497 [-] on Build GRCh38 |
123 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105373085 |
LOC105373085 |
|
-364.4kb
|
Sense |
Chr. - 49047480 - 49052405 on Build GRCh38 |
Downstream |
microRNA 3667 |
MIR3667 |
|
102.2kb
|
Sense |
Chr. - 49543393 - 49543466 on Build GRCh38 |
View Full Product Details