Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NDUFA5
- Entrez Gene ID
-
4698
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A5
- Gene Aliases
-
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13
- Chromosome Location
-
Chr.7: 123536997 - 123601651 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111926.1 |
|
3 - 4 |
228 |
197 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
IQ motif and ubiquitin domain containing |
IQUB |
|
-36.3kb
|
Sense |
Chr. - 123452181 - 123535077 on Build GRCh38 |
Downstream |
uncharacterized LOC102724555 |
LOC102724555 |
|
29.2kb
|
Sense |
Chr. - 123614161 - 123624957 on Build GRCh38 |
View Full Product Details