Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLUD1P7
Entrez Gene ID
414212
Gene Name
glutamate dehydrogenase 1 pseudogene 7
Gene Aliases
GLUD1P2, GLUDP2, GLUDP7, bA342C24.3
Chromosome Location
Chr.10: 46759023 - 46786770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_111968.1 1 - 2 68 117
GenBank mRNA BX281697.1 - 1 - 2 68 117


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC102724643 LOC102724643 -23.6kb Sense Chr. - 47587185 - 47619491 on Build GRCh38
Downstream FERM and PDZ domain containing 2B, pseudogene FRMPD2B FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2 89.9kb Anti-sense Chr. - 46876674 - 46895326 on Build GRCh38

View Full Product Details