Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ATP11A-AS1
- Entrez Gene ID
-
100874205
- Gene Name
- ATP11A antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 112745449 - 112754731 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046661.1 |
|
1 - 2 |
236 |
89 |
GenBank mRNA |
BC041346.1 |
- |
1 - 2 |
200 |
89 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI465087.1 |
- |
1 - 2 |
200 |
|
Chr. 13 - 112746851 - 112754693 [-] on Build GRCh38 |
89 |
GenBankmRNA |
DB095404.1 |
- |
1 - 2 |
217 |
|
Chr. 13 - 112747101 - 112754712 [-] on Build GRCh38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATP11A upstream neighbor |
ATP11AUN |
C13orf35, SMABLO1 |
-61.0kb
|
Anti-sense |
Chr. - 112647044 - 112684497 on Build GRCh38 |
Downstream |
MCF.2 cell line derived transforming sequence like |
MCF2L |
ARHGEF14, DBS, OST |
139.6kb
|
Anti-sense |
Chr. - 112894378 - 113099739 on Build GRCh38 |
View Full Product Details