The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024144.2 | 1 - 2 | 47 | 73 | |
NR_073567.1 | 1 - 2 | 47 | 73 | ||
NR_073568.1 | 1 - 2 | 47 | 73 | ||
GenBank mRNA | EU789529.1 | - | 1 - 2 | 47 | 73 |
EU789535.1 | - | 1 - 2 | 46 | 73 | |
EU789536.1 | - | 1 - 2 | 46 | 73 | |
EU789544.1 | - | 1 - 2 | 46 | 73 | |
EU789545.1 | - | 1 - 2 | 46 | 73 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | TatD DNase domain containing 2 | TATDN2 | Sense | Chr. - 10248493 - 10281222 on Build GRCh38 | ||
Downstream | long intergenic non-protein coding RNA 852 | LINC00852 | C3orf42, GHRL-AS2, GHRLOS2, NAG73 | 816.0 | Sense | Chr. - 10284419 - 10285746 on Build GRCh38 |