Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
Chromosome Location
Chr.3: 10280952 - 10293449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024144.2 2 - 3 131 83
NR_024145.2 3 - 4 288 83
NR_073566.1 3 - 4 227 83
NR_073567.1 2 - 3 131 83
NR_073568.1 2 - 3 131 83
GenBank mRNA EU789528.1 - 3 - 4 288 83
EU789529.1 - 2 - 3 131 83
EU789535.1 - 2 - 3 130 83
EU789536.1 - 2 - 3 130 83
EU789537.1 - 3 - 4 287 83
EU789543.1 - 3 - 4 226 83
EU789544.1 - 2 - 3 130 83
EU789545.1 - 2 - 3 130 83


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 852 LINC00852 C3orf42, GHRL-AS2, GHRLOS2, NAG73 Sense Chr. - 10284419 - 10285746 on Build GRCh38
Downstream SEC13 homolog, nuclear pore and COPII coat complex component SEC13 D3S1231E, SEC13L1, SEC13R, npp-20 15.1kb Anti-sense Chr. - 10300929 - 10321188 on Build GRCh38

View Full Product Details