Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC00408
- Entrez Gene ID
-
100652856
- Gene Name
- long intergenic non-protein coding RNA 408
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 18905439 - 18926741 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104118.1 |
|
3 - 4 |
377 |
68 |
GenBank mRNA |
BC035261.1 |
- |
3 - 4 |
378 |
68 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG722014.1 |
- |
3 - 4 |
383 |
|
Chr. 13 - 18905433 - 18926484 [+] on Build GRCh38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ankyrin repeat domain 20 family member A9, pseudogene |
ANKRD20A9P |
|
-34.5kb
|
Anti-sense |
Chr. - 18834403 - 18871969 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 442 |
LINC00442 |
|
100.5kb
|
Sense |
Chr. - 19008259 - 19012634 on Build GRCh38 |
View Full Product Details