Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC101929279
- Entrez Gene ID
-
101929279
- Gene Name
- uncharacterized LOC101929279
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 30211852 - 30232690 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120653.1 |
|
3 - 3 |
1020 |
115 |
GenBank mRNA |
BC043271.2 |
- |
3 - 3 |
1020 |
115 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
KIAA1462 |
KIAA1462 |
JCAD |
-115.4kb
|
Anti-sense |
Chr. - 30012800 - 30069816 on Build GRCh38 |
Downstream |
mitochondrial poly(A) polymerase |
MTPAP |
PAPD1, SPAX4 |
77.1kb
|
Anti-sense |
Chr. - 30309801 - 30349338 on Build GRCh38 |
View Full Product Details