Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORA68
- Entrez Gene ID
-
26780
- Gene Name
- small nucleolar RNA, H/ACA box 68
- Gene Aliases
-
RNU68, SNORA68A, U68
- Chromosome Location
-
Chr.19: 17862588 - 17862720 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_000012.1 |
|
1 - 1 |
80 |
92 |
GenBank mRNA |
Y11162.1 |
- |
1 - 1 |
80 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ribosomal protein L18a |
RPL18A |
L18A |
|
Sense |
Chr. - 17859878 - 17863324 on Build GRCh38 |
Downstream |
solute carrier family 5 member 5 |
SLC5A5 |
NIS, TDH1 |
8.7kb
|
Sense |
Chr. - 17871394 - 17895175 on Build GRCh38 |
View Full Product Details