Product Details
- Assay Reference Genome
Location
Chr.10:93780744 on build GRCh38
- Cytoband
- 10q23.33
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs76315560] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 9211
- Gene Symbol
- LGI1
- Gene Name
- leucine rich glioma inactivated 1
- Gene Aliases
- ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
- Location
- Chr.10:93757770-93798174 on build GRCh38
- Assay Gene Location
- Within Intron 4
View Full Product Details