Product Details

Assay Reference Genome
Location

Chr.20:56435165 on build GRCh38
Cytoband
20q13.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
57091
Gene Symbol
CASS4
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
Location
Chr.20:56411550-56460240 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CASS4 NM_001164114.1 NP_001157586.1
NM_001164115.1 NP_001157587.1
NM_001164116.1 NP_001157588.1
NM_020356.3 NP_065089.2
XM_006723831.3 XP_006723894.1
AJ276678.1 CAC00655.1
AK027760.1 BAB55351.1
AK295613.1
AK308057.1
AK314653.1
BC027951.1 AAH27951.1
BX280195.1
DA584687.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv787e214 Chr20:56429038 - 56481532 on Build GRCh38 Gain CASS4 RTFDC1
esv3646189 Chr20:56430399 - 56481532 on Build GRCh38 Loss CASS4 RTFDC1
nsv525425 Chr20:56403979 - 56897294 on Build GRCh38 Gain FAM209A CSTF1 LOC105372682 CASS4 TFAP2C RTFDC1 GCNT7 FAM209B
nsv834013 Chr20:56307279 - 56476126 on Build GRCh38 Gain FAM210B CSTF1 CASS4 RTFDC1 AURKA
esv3646191 Chr20:56431805 - 56436322 on Build GRCh38 Loss CASS4

View Full Product Details