Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
HNF4A | NM_001030003.2 | NP_001025174.1 | ||
NM_001030004.2 | NP_001025175.1 | |||
NM_001287182.1 | NP_001274111.1 | |||
NM_001287183.1 | NP_001274112.1 | |||
NM_001287184.1 | NP_001274113.1 | |||
NM_175914.4 | NP_787110.2 | |||
AK096973.1 | 1 | 762 | ||
AY680696.1 | AAT91237.1 | |||
AY680697.1 | AAT91238.1 | |||
AY680698.1 | AAT91239.1 | |||
FJ608822.1 | ||||
HQ692868.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057847 | Chr20:42652464 - 44431352 on Build GRCh38 | Loss | HNF4A OSER1-AS1 R3HDML L3MBTL1 SGK2 LOC101927159 LOC101927200 GTSF1L FITM2 LOC101927138 LOC105372626 MYBL2 GDAP1L1 SRSF6 MIR3646 TOX2 HNF4A-AS1 JPH2 OSER1 IFT52 PTPRT |
nsv586065 | Chr20:44261867 - 44505244 on Build GRCh38 | Gain | HNF4A R3HDML TTPAL GDAP1L1 SERINC3 MIR3646 HNF4A-AS1 LINC01620 FITM2 LINC01430 |