Product Details

Assay Reference Genome
Location

Chr.5:151021485 on build GRCh38
Cytoband
5q33.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2878
Gene Symbol
GPX3
Gene Name
glutathione peroxidase 3
Gene Aliases
GPx-P, GSHPx-3, GSHPx-P
Location
Chr.5:151020438-151028993 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GPX3 NM_002084.3 NP_002075.2
AF217787.1 AAF43005.1
AK310242.1
BC013601.1 AAH13601.1
BC035841.1 AAH35841.1
BC050378.1 AAH50378.2
BC066949.1 AAH66949.1
BI522602.1
D00632.1 BAA00525.1
X58295.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv526224 Chr5:151003841 - 151070675 on Build GRCh38 Gain TNIP1 GPX3

View Full Product Details