Product Details

Assay Reference Genome
Location

Chr.11:112977048 on build GRCh38
Cytoband
11q23.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4684
Gene Symbol
NCAM1
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
Location
Chr.11:112961247-113278436 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NCAM1 NM_000615.6 NP_000606.3
NM_001076682.3 NP_001070150.1
NM_001242607.1 NP_001229536.1
NM_001242608.1 NP_001229537.1
NM_181351.4 NP_851996.2
AB209443.1 BAD92680.1
AF005070.1 AAB61615.1
AK292453.1
AK314589.1
BC029119.1
BC047244.1 AAH47244.1
DA231540.1
DA337649.1
S71824.1 AAB31836.1
U63041.1 AAB04558.1
X16841.1 CAA34739.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv832271 Chr11:112972081 - 113007925 on Build GRCh38 Gain NCAM1
dgv1279n100 Chr11:112966837 - 112987859 on Build GRCh38 Loss NCAM1

View Full Product Details