Product Details

Assay Reference Genome
Location

Chr.17:7695484 on build GRCh38
Cytoband
17p13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55135
Gene Symbol
WRAP53
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
Location
Chr.17:7686071-7703502 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRAP53 NM_001143990.1 NP_001137462.1
NM_001143991.1 NP_001137463.1
NM_001143992.1 NP_001137464.1
NM_018081.2 NP_060551.2
XM_011523952.2 XP_011522254.1
XM_017024812.1 XP_016880301.1
AK001247.1 BAA91579.1
AK056669.1
AK092370.1
AK225273.1
AK225411.1
AK303246.1
AY766322.1
BC002336.2 AAH02336.1
DQ431240.1
DQ431241.1

View Full Product Details