Product Details

Assay Reference Genome
Location

Chr.1:11955265 on build GRCh38
Cytoband
1p36.22
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5351
Gene Symbol
PLOD1
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
Location
Chr.1:11934667-11975542 on build GRCh38
Assay Gene Location
Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PLOD1 NM_000302.3 NP_000293.2
NM_001316320.1 NP_001303249.1
AK294688.1
AK295254.1
AK299150.1
AK312243.1
AK316285.1
BC016657.2 AAH16657.1
M98252.1

View Full Product Details