Product Details

Assay Reference Genome
Location

Chr.9:127472828 on build GRCh38
Cytoband
9q33.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
90678
Gene Symbol
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Location
Chr.9:127451486-127503501 on build GRCh38
Assay Gene Location
Within Intron 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LRSAM1 NM_001005373.3 NP_001005373.1
NM_001005374.3 NP_001005374.1
NM_001190723.2 NP_001177652.1
NM_138361.5 NP_612370.3
XM_006717316.3 XP_006717379.1
XM_017015283.1 XP_016870772.1
XM_017015284.1 XP_016870773.1
AK056203.1 BAB71119.1
AK091589.1 BAC03703.1
AK226106.1
AY358830.1 AAQ89189.1
BC009239.2 AAH09239.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv466569 Chr9:127470144 - 127512011 on Build GRCh38 Loss LRSAM1 FAM129B

View Full Product Details