Gene Transcripts

Gene Symbol
Srl
Entrez Gene ID
106393
Gene Name
sarcalumenin
Gene Aliases
9830004M20Rik, AW108387, sar
Chromosome Location
Chr. 16: 4480212 - 4523627 on Build GRCm38
UniGene ID
Mm.35811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175347.4 NP_780556.2 2 583
GenBank mRNA AB113383.1 - 2 583
BC145687.1 - 2 605 40130990

View Full Product Details