Gene Transcripts

Gene Symbol
Exoc6
Entrez Gene ID
107371
Gene Name
exocyst complex component 6
Gene Aliases
4833405E05Rik, AW413330, C430002C19, Sec15, Sec15l1, hbd, msec15
Chromosome Location
Chr. 19: 37520874 - 37684052 on Build GRCm38
UniGene ID
Mm.24865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175353.2 NP_780562.2 6 594
GenBank mRNA AK082881.1 - 6 573
AK151524.1 - 6 595
AK151838.1 - 6 595
BC026859.1 - 6 547 2631201
CD806857.1 - 1 92

View Full Product Details