Gene Transcripts
- Gene Symbol
- Tm6sf1
- Entrez Gene ID
- 107769
- Gene Name
- transmembrane 6 superfamily member 1
- Gene Aliases
- AI428514, BC023123, C630016D09Rik
- Chromosome Location
- Chr. 7: 81859001 - 81884079 on Build GRCm38
- UniGene ID
- Mm.221412
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145375.4 |
NP_663350.2
|
1, 2 |
542 |
|
GenBank mRNA |
AK046714.1 |
- |
1, 2 |
443 |
|
AK083132.1 |
- |
1, 2 |
545 |
|
AK135100.1 |
- |
1, 2 |
524 |
|
AK162572.1 |
- |
1, 2 |
394 |
|
AK163621.1 |
- |
1, 2 |
467 |
|
AK172109.1 |
- |
1, 2 |
458 |
|
AK172145.1 |
- |
1, 2 |
443 |
|
BB653227.1 |
- |
1, 2 |
544 |
|
BC023123.1 |
- |
1, 2 |
424 |
3995280 |
View Full Product Details