Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102455.1 | NP_001095925.1 | 16 | 2116 | |
NM_001102456.1 | NP_001095926.1 | 16 | 1984 | ||
NM_009691.2 | NP_033821.1 | 15 | 1948 | ||
GenBank mRNA | AK088391.1 | - | 3 | 339 | |
AK149977.1 | - | 16 | 2117 | ||
AK150020.1 | - | 16 | 2117 | ||
AK150478.1 | - | 8 | 848 | ||
AK151708.1 | - | 8 | 848 | ||
AK152787.1 | - | 8 | 848 | ||
AK153087.1 | - | 8 | 848 | ||
BC026491.1 | - | 6 | 400 | ||
BC051999.1 | - | 16 | 1962 | 5716148 | |
BC052396.1 | - | 15 | 1920 | 5719918 | |
M97216.1 | - | 11 | 1308 | ||
U15571.1 | - | 16 | 2092 | ||
Z22592.1 | - | 15 | 1948 | ||
Z27070.1 | - | 15 | 1946 |