Gene Transcripts

Gene Symbol
Serping1
Entrez Gene ID
12258
Gene Name
serine (or cysteine) peptidase inhibitor, clade G, member 1
Gene Aliases
C1INH, C1nh
Chromosome Location
Chr. 2: 84765360 - 84775445 on Build GRCm38
UniGene ID
Mm.38888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009776.3 NP_033906.2 8 1561
GenBank mRNA AA237518.1 - 2 273
AF010254.1 - 8 1536
AK144292.1 - 7 1521
AK165744.1 - 7 1507
AK167585.1 - 7 1498
BC002026.1 - 7 1492 3485810
BQ031701.1 - 1 273
Y10386.1 - 8 1518

View Full Product Details