Gene Transcripts
- Gene Symbol
- Cacnb4
- Entrez Gene ID
- 12298
- Gene Name
- calcium channel, voltage-dependent, beta 4 subunit
- Gene Aliases
- 3110038O15Rik, Cchb4, lh
- Chromosome Location
- Chr. 2: 52428320 - 52676609 on Build GRCm38
- UniGene ID
- Mm.330223
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037099.2 |
NP_001032176.1
|
12, 13 |
1395 |
|
NM_001285426.1 |
NP_001272355.1
|
12, 13 |
1179 |
|
NM_001285427.1 |
NP_001272356.1
|
12, 13 |
1475 |
|
NM_001285428.1 |
NP_001272357.1
|
11, 12 |
1104 |
|
NM_146123.3 |
NP_666235.1
|
11, 12 |
1315 |
|
GenBank mRNA |
AF039417.1 |
- |
2, 3 |
172 |
|
AK032311.1 |
- |
12, 13 |
1618 |
|
AK038633.1 |
- |
12, 13 |
1431 |
|
AK043850.1 |
- |
11, 12 |
1258 |
|
AK079616.1 |
- |
11, 12 |
1265 |
|
AK147338.1 |
- |
12, 13 |
1369 |
|
AK158606.1 |
- |
11, 12 |
1106 |
|
BC026479.1 |
- |
11, 12 |
1101 |
4501980 |
View Full Product Details