Gene Transcripts

Gene Symbol
Cyp17a1
Entrez Gene ID
13074
Gene Name
cytochrome P450, family 17, subfamily a, polypeptide 1
Gene Aliases
Cyp17, p450c17
Chromosome Location
Chr. 19: 46667165 - 46673000 on Build GRCm38
UniGene ID
Mm.1262
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007809.3 NP_031835.3 5 1040
GenBank mRNA AK132830.1 - 5 934
AK134376.1 - 5 934
BC064793.1 - 5 1014 30436278
M64863.1 - 5 913

View Full Product Details