Gene Transcripts

Gene Symbol
Dnah11
Entrez Gene ID
13411
Gene Name
dynein, axonemal, heavy chain 11
Gene Aliases
Dnahc11, b2b1203Clo, b2b1279Clo, b2b1289Clo, b2b1727Clo, b2b598Clo, iv, lrd
Chromosome Location
Chr. 12: 117877982 - 118199043 on Build GRCm38
UniGene ID
Mm.390540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010060.3 NP_034190.3 62 10181
GenBank mRNA AF183144.1 - 62 10181
BC058762.1 - 29 4259

View Full Product Details