Gene Transcripts

Gene Symbol
Evi5
Entrez Gene ID
14020
Gene Name
ecotropic viral integration site 5
Gene Aliases
NB4S
Chromosome Location
Chr. 5: 107744795 - 107875107 on Build GRCm38
UniGene ID
Mm.35796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007964.2 NP_031990.2 13, 14 1695
GenBank mRNA AK134294.1 - 15, 16 2076
AK136326.1 - 3, 4 476
CV557033.1 - 4, 5 381
U53586.1 - 13, 14 1695

View Full Product Details